N,N,N',N'-Tetrakis[4-(diisobutylamino)phenyl]-1,4-phenylenediamine 98%

Supplier: Ambeed
Warning

Synonyms: N1,N1'-(1,4-Phenylene)bis(N1-(4-(diisobutylamino)phenyl)-N4,N4-diisobutylbenzene-1,4-diamine), N1,N1'-(1,4-Phenylene)bis[N1-[4-(diisobutylamino)phenyl]-N4,N4-diisobutylbenzene-1,4-diamine]

A855032-1G A855032-250MG A855032-5G
76993-618EA 14 USD
76993-618 76993-620 76993-622
N,N,N',N'-Tetrakis[4-(diisobutylamino)phenyl]-1,4-phenylenediamine 98%
N,N,N',N'-Tetrakis[4-(diisobutylamino)phenyl]-1,4-phenylenediamine

Precaución: for Research Use Only

Formula: C₆₂H₉₂N₆
MDL Number: MFCD22207043
CAS Number: 485831-34-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR