2,6-Difluoro-3-propoxyphenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (2,6-Difluoro-3-propoxyphenyl)boronic acid

A853951-1G A853951-250MG A853951-5G
77019-706EA 68.96 USD
77019-706 77019-708 77019-710
2,6-Difluoro-3-propoxyphenylboronic acid 97%
2,6-Difluoro-3-propoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BF₂O₃
MW: 215.99 g/mol
MDL Number: MFCD07369752
CAS Number: 849062-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR