tert-Butyl-2-(piperidin-4-yl)acetate oxalate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2-(piperidin-4-yl)acetate oxalate

A853877-1G A853877-250MG
77019-702EA 125.19 USD
77019-702 77019-704
tert-Butyl-2-(piperidin-4-yl)acetate oxalate 95%
tert-Butyl-2-(piperidin-4-yl)acetate oxalate

Precaución: for Research Use Only

Formula: C₁₃H₂₃NO₆
MW: 289.33 g/mol
MDL Number: MFCD16659936
CAS Number: 1360438-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR