2-Methoxy-6-methylpyridine-3-boronic acid 98%
Supplier: Ambeed
|
Warning
|
A853791-250MG
A853791-1G
A853791-5G
76993-608EA
63
USD
76993-608
77891-610
77891-611
2-Methoxy-6-methylpyridine-3-boronic acid 98%
2-Methoxy-6-methylpyridine-3-boronic acid
Precaución: for Research Use Only
Formula:
C₇H₁₀BNO₃ |
MDL Number:
MFCD07374993 CAS Number: 1000802-75-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...