2,3-O-Isopropylidene-D-glyceraldehyde 95%

Supplier: Ambeed
Danger

Synonyms: (4R)-2,2-dimethyl-1,3-dioxolane-4-carbaldehyde, (R)-(+)-2,2-Dimethyl-1,3-dioxolane-4-carboxaldehyde, (R)-2,2-dimethyl-1,3-dioxolane-4-carbaldehyde

A852953-25G A852953-5G
77006-844EA 16.98 USD
77006-844 77006-846
2,3-O-Isopropylidene-D-glyceraldehyde 95%
2,3-O-Isopropylidene-D-glyceraldehyde

Precaución: for Research Use Only

Formula: C₆H₁₀O₃
MW: 130.143 g/mol
MDL Number: MFCD00269682
CAS Number: 15186-48-8
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR