6-Fluoro-3,4-dihydro-2H-isoquinolin-1-one 96%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Fluoro-3,4-dihydroisoquinolin-1(2H)-one
A852733-250MG
A852733-100MG
77019-700EA
50.93
USD
77019-700
77684-814
6-Fluoro-3,4-dihydro-2H-isoquinolin-1-one 96%, Ambeed.Inc
6-Fluoro-3,4-dihydro-2H-isoquinolin-1-one
Precaución: For research use only.
Formula:
C₉H₈FNO MW: 165.164 g/mol |
MDL Number:
MFCD04114864 CAS Number: 214045-84-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...