(2-(Methylthio)pyrimidin-4-yl)methanamine 95%

Supplier: Ambeed
Warning

A852595-1G A852595-250MG
77019-696EA 485.9 USD
77019-696 77019-698
(2-(Methylthio)pyrimidin-4-yl)methanamine 95%
(2-(Methylthio)pyrimidin-4-yl)methanamine

Precaución: for Research Use Only

Formula: C₆H₉N₃S
MW: 155.22 g/mol
MDL Number: MFCD19981683
CAS Number: 1312095-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR