5-Acetamido-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodoisophthalamide 98%

Supplier: Ambeed
Warning

A852530-100G A852530-25G A852530-500G A852530-5G
77006-836EA 46 USD
77006-836 77006-838 77006-840 77006-842
5-Acetamido-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodoisophthalamide 98%
5-Acetamido-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodoisophthalamide

Precaución: for Research Use Only

Formula: C₁₆H₂₀I₃N₃O₇
MDL Number: MFCD08063354
CAS Number: 31127-80-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR