5-Bromo-4-methylthiophene-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A852485-1G A852485-5G
76993-604EA 61.54 USD
76993-604 76993-606
5-Bromo-4-methylthiophene-2-carboxylic acid 98%
5-Bromo-4-methylthiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₅BrO₂S
MW: 221.07 g/mol
MDL Number: MFCD11110671
CAS Number: 54796-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR