Ethyl-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate 95+%

Supplier: Ambeed
Warning

Synonyms: Ethyl 6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate

A852240-1G A852240-250MG A852240-5G
77006-830EA 50 USD
77006-830 77006-832 77006-834
Ethyl-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate 95+%
Ethyl 6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₉F₃N₂O₂
MW: 258.20 g/mol
MDL Number: MFCD11100212
CAS Number: 860457-99-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR