Fmoc-O-phospho-L-tyrosine 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-O-Phospho-Tyrosine, Fmoc-Tyr(H₂PO₃)-OH, Fmoc-Tyr(H2PO3)-OH, Fmoc-Tyr(PO₃H₂)-OH

A852141-1G A852141-250MG A852141-5G A852141-25G
77006-824EA 96.55 USD
77006-824 77006-826 77006-828 77674-262
Fmoc-O-phospho-L-tyrosine 98%
Fmoc-O-phospho-L-tyrosine

Precaución: for Research Use Only

Formula: C₂₄H₂₂NO₈P
MW: 483.413 g/mol
MDL Number: MFCD00235902
CAS Number: 147762-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR