tert-Butyl naphthalen-1-ylcarbamate 96%

Supplier: Ambeed
Warning

A852107-1G A852107-5G
77036-684EA 29.71 USD
77036-684 77036-686
tert-Butyl naphthalen-1-ylcarbamate 96%
tert-Butyl naphthalen-1-ylcarbamate

Precaución: for Research Use Only

Formula: C₁₅H₁₇NO₂
MW: 243.30 g/mol
MDL Number: MFCD07127697
CAS Number: 72594-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR