Methyl 2-benzyl-5,6-dihydroxypyrimidine-4-carboxylate 95%

Supplier: Ambeed
Warning

A851948-100MG A851948-1G A851948-250MG
77019-690EA 85.94 USD
77019-690 77019-692 77019-694
Methyl 2-benzyl-5,6-dihydroxypyrimidine-4-carboxylate 95%
Methyl 2-benzyl-5,6-dihydroxypyrimidine-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₂N₂O₄
MW: 260.25 g/mol
MDL Number: MFCD09031861
CAS Number: 519032-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR