2,4-Dichloro-7-fluoro-8-methylquinazoline 95%

Supplier: Ambeed
Danger

A849632-100MG A849632-1G A849632-250MG A849632-5G
77019-682EA 45.62 USD
77019-682 77019-684 77019-686 77019-688
2,4-Dichloro-7-fluoro-8-methylquinazoline 95%
2,4-Dichloro-7-fluoro-8-methylquinazoline

Precaución: for Research Use Only

Formula: C₉H₅Cl₂FN₂
MW: 231.05 g/mol
MDL Number: MFCD27997107
CAS Number: 1039736-73-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR