1-Methyl-3-nitro-1H-pyrazole 98%
Supplier: Ambeed
|
Warning
|
A849463-1G
A849463-250MG
A849463-5G
77036-674EA
58.35
USD
77036-674
77036-676
77036-678
1-Methyl-3-nitro-1H-pyrazole 98%
1-Methyl-3-nitro-1H-pyrazole
Precaución: for Research Use Only
Formula:
C₄H₅N₃O₂ MW: 127.101 g/mol |
MDL Number:
MFCD03444368 CAS Number: 54210-32-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...