2-Methyl-2H-indazole-7-carboxylic acid methyl ester 98%

Supplier: Ambeed
Danger

A849326-1G A849326-250MG A849326-5G
77036-668EA 15 USD
77036-668 77036-670 77036-672
2-Methyl-2H-indazole-7-carboxylic acid methyl ester 98%
2-Methyl-2H-indazole-7-carboxylic acid methyl ester

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂
MDL Number: MFCD11109408
CAS Number: 1092351-88-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR