(2S)-Morpholine-2-carboxylic acid hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: (S)-Morpholine-2-carboxylic acid hydrochloride

A848763-100MG A848763-250MG
77019-674EA 27.59 USD
77019-674 77019-676
(2S)-Morpholine-2-carboxylic acid hydrochloride 95%
(2S)-Morpholine-2-carboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₅H₉NO₃·ClH
MW: 167.591 g/mol
MDL Number: MFCD13186759
CAS Number: 1439373-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR