4-(1-Azetidinylsulfonyl)benzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-(Azetidin-1-ylsulphonyl)phenyl)boronic acid

A848136-100MG A848136-1G A848136-250MG A848136-5G
77019-666EA 28 USD
77019-666 77019-668 77019-670 77019-672
4-(1-Azetidinylsulfonyl)benzeneboronic acid 98%
4-(1-Azetidinylsulfonyl)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₂BNO₄S
MDL Number: MFCD07363756
CAS Number: 871329-68-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR