Eliglustat 98%

Supplier: Ambeed
Warning

Synonyms: Genz99067, N-((1R,2R)-1-(2,3-dihydrobenzo[b][1,4]dioxin-6-yl)-1-hydroxy-3-(pyrrolidin-1-yl)propan-2-yl)octanamide hemi((2R,3R)-2,3-dihydroxysuccinate)

A848004-10MG A848004-1MG A848004-5MG
76993-584EA 44.56 USD
76993-584 76993-586 76993-588
Eliglustat 98%
Eliglustat

Precaución: for Research Use Only

Formula: C₂₃H₃₆N₂O
MW: 404.54 g/mol
MDL Number: MFCD19443735
CAS Number: 491833-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR