Boc-D-Thr(Bzl)-OH 98%

Supplier: Ambeed
Warning

Synonyms: N-Boc-O-benzyl-D-threonine, (2R,3S)-3-(Benzyloxy)-2-((tert-butoxycarbonyl)amino)butanoic acid

A847797-10G A847797-1G A847797-250MG
77036-662EA 142.17 USD
77036-662 77036-664 77036-666
Boc-D-Thr(Bzl)-OH 98%
Boc-D-Thr(Bzl)-OH

Precaución: for Research Use Only

Formula: C₁₆H₂₃NO₅
MW: 309.362 g/mol
MDL Number: MFCD00038247
CAS Number: 69355-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR