3-Iodo-4-(trifluoromethoxy)benzoic acid 95%

Supplier: Ambeed
Warning

A847717-1G A847717-250MG
77036-658EA 218 USD
77036-658 77036-660
3-Iodo-4-(trifluoromethoxy)benzoic acid 95%
3-Iodo-4-(trifluoromethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₄F₃IO₃
MDL Number: MFCD08459287
CAS Number: 1110709-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR