2-Bromo-5-hexylthiophene 98%

Supplier: Ambeed
Danger

Synonyms: 5-Bromo-2-hexylthiophene

A847688-1G A847688-250MG A847688-25G A847688-5G
76993-576EA 25.47 USD
76993-576 76993-578 76993-580 76993-582
2-Bromo-5-hexylthiophene 98%
2-Bromo-5-hexylthiophene

Precaución: for Research Use Only

Formula: C₁₀H₁₅BrS
MW: 247.19 g/mol
MDL Number: MFCD10000906
CAS Number: 211737-28-9
UN: 2922
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR