2-(Methylamino)-5-pyrimidineboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methylaminopyrimidine-5-boronic acid, pinacol ester, N-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine

A847562-100MG A847562-1G A847562-250MG A847562-5G
76993-570EA 37 USD
76993-570 76993-572 76993-574 77891-601
2-(Methylamino)-5-pyrimidineboronic acid pinacol ester 98%
2-(Methylamino)-5-pyrimidineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₈BN₃O₂
MDL Number: MFCD11878345
CAS Number: 904326-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR