3-Aminoadamantane-1-carboxylic acid hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A847500-250MG
A847500-5G
76993-564EA
14.86
USD
76993-564
76993-566
76993-568
3-Aminoadamantane-1-carboxylic acid hydrochloride 95%
3-Aminoadamantane-1-carboxylic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₁₁H₁₈ClNO₂ |
MDL Number:
MFCD01825827 CAS Number: 6240-01-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...