2-(1,3,5-Trimethyl-1H-pyrazol-4-yl)ethanol 95%

Supplier: Ambeed
Warning

A846657-100MG A846657-250MG
77006-802EA 53.05 USD
77006-802 77006-804
2-(1,3,5-Trimethyl-1H-pyrazol-4-yl)ethanol 95%
2-(1,3,5-Trimethyl-1H-pyrazol-4-yl)ethanol

Precaución: for Research Use Only

Formula: C₈H₁₄N₂O
MW: 154.21 g/mol
MDL Number: MFCD08060042
CAS Number: 1007462-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR