3-Chloro-4-(3'-chlorobenzyloxy)phenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (3-Chloro-4-((3-chlorobenzyl)oxy)phenyl)boronic acid

A846609-1G A846609-250MG A846609-5G
76993-556EA 97.61 USD
76993-556 76993-558 76993-560
3-Chloro-4-(3'-chlorobenzyloxy)phenylboronic acid 95%
3-Chloro-4-(3'-chlorobenzyloxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁BCl₂O₃
MW: 296.94 g/mol
MDL Number: MFCD06411318
CAS Number: 849062-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR