Dodecyltrimethylammonium bromide 98%

Supplier: Ambeed
Danger

A845744-25G A845744-100G
77006-800EA 20 USD
77006-800 77603-144
Dodecyltrimethylammonium bromide 98%
Dodecyltrimethylammonium bromide

Precaución: for Research Use Only

Formula: C₁₅H₃₄BrN
MDL Number: MFCD00011767
CAS Number: 1119-94-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR