Boc-n-Me-abz-OH 98%

Supplier: Ambeed
Warning

Synonyms: 2-((tert-Butoxycarbonyl)(methyl)amino)benzoic acid, 2-(N-Boc-N-methylamino)benzoic acid

A845645-1G A845645-250MG A845645-5G
76993-550EA 109 USD
76993-550 76993-552 76993-554
Boc-n-Me-abz-OH 98%
Boc-n-Me-abz-OH

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₄
MW: 251.282 g/mol
MDL Number: MFCD00237567
CAS Number: 141871-02-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR