1-(Cbz-amino)cyclohexanecarboxylic Acid 98%
Supplier: Ambeed
|
Warning
|
A845528-100MG
A845528-1G
A845528-250MG
A845528-5G
77006-792EA
5
USD
77006-792
77006-794
77006-796
77006-798
1-(Cbz-amino)cyclohexanecarboxylic Acid 98%
1-(Cbz-amino)cyclohexanecarboxylic Acid
Precaución: for Research Use Only
Formula:
C₁₅H₁₉NO₄ |
MDL Number:
MFCD02094521 CAS Number: 17191-43-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...