4-(Diphenylphosphino)benzaldehyde 98%
Supplier: Ambeed
|
Warning
|
A844008-100MG
A844008-1G
A844008-250MG
76993-540EA
42.44
USD
76993-540
76993-542
77684-808
4-(Diphenylphosphino)benzaldehyde 98%
4-(Diphenylphosphino)benzaldehyde
Precaución: For research use only.
Formula:
C₁₉H₁₅OP MW: 290.29 g/mol |
MDL Number:
MFCD26743564 CAS Number: 5068-18-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...