2-(3-(Trifluoromethyl)pyridin-2-yl)acetic acid 95%

Supplier: Ambeed
Warning

A843982-250MG A843982-100MG
77036-656EA 270 USD
77036-656 77873-309
2-(3-(Trifluoromethyl)pyridin-2-yl)acetic acid 95%
2-(3-(Trifluoromethyl)pyridin-2-yl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₆F₃NO₂
MDL Number: MFCD09924925
CAS Number: 1000521-27-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR