2-Fluoro-6-formylbenzonitrile 95%

Supplier: Ambeed
Warning

A842810-100MG A842810-1G A842810-250MG
77019-638EA 36 USD
77019-638 77019-640 77019-642
2-Fluoro-6-formylbenzonitrile 95%
2-Fluoro-6-formylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₄FNO
MW: 149.12 g/mol
MDL Number: MFCD18394004
CAS Number: 1261823-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR