N-Carbobenzoxy-D-leucine dicyclohexylammonium salt 97%

Supplier: Ambeed
Warning

Synonyms: N-Cbz-D-leucine Dicyclohexylammonium Salt, Z-D-Leu-OH.DCHA

A841832-100G A841832-25G A841832-5G
76993-532EA 53 USD
76993-532 76993-534 76993-536
N-Carbobenzoxy-D-leucine dicyclohexylammonium salt 97%
N-Carbobenzoxy-D-leucine dicyclohexylammonium salt

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₄
MW: 446.63 g/mol
MDL Number: MFCD10566885
CAS Number: 7662-58-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR