4,5-Diphenyloxazole 95%

Supplier: Ambeed
Warning

A841494-1G A841494-100MG A841494-250MG
76993-530EA 616 USD
76993-530 77891-590 77891-591
4,5-Diphenyloxazole 95%
4,5-Diphenyloxazole

Precaución: for Research Use Only

Formula: C₁₅H₁₁NO
MDL Number: MFCD16990157
CAS Number: 4675-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR