6-(tert-Butoxycarbonylamino)-1-hexanol 97%

Supplier: Ambeed
Warning

Synonyms: N-(6-Hydroxyhexyl)carbamic Acid tert-Butyl Ester, Boc-Ahx-ol, tert-Butyl N-(6-Hydroxyhexyl)carbamate, N-Boc-6-aminohexanol, N-Boc-6-hydroxyhexylamine

A840492-1G A840492-250MG A840492-25G A840492-10G A840492-100G A840492-5G
76993-524EA 47.75 USD
76993-524 76993-526 77573-670 77614-508 77684-802 77684-803
6-(tert-Butoxycarbonylamino)-1-hexanol 97%
6-(tert-Butoxycarbonylamino)-1-hexanol

Precaución: For research use only.

Formula: C₁₁H₂₃NO₃
MW: 217.31 g/mol
MDL Number: MFCD01862954
CAS Number: 75937-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR