3-Bromo-5-chloroanthranilic acid 95%

Supplier: Ambeed
Warning

A840218-1G A840218-250MG A840218-5G A840218-25G A840218-100G
77036-630EA 22 USD
77036-630 77036-632 77036-634 77573-668 77873-306
3-Bromo-5-chloroanthranilic acid 95%
3-Bromo-5-chloroanthranilic acid

Precaución: for Research Use Only

Formula: C₇H₅BrClNO₂
MDL Number: MFCD04035658
CAS Number: 41198-02-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR