3-Bromodibenzothiophene 98%

Supplier: Ambeed
Warning

A840198-10G A840198-1G A840198-250MG A840198-5G
77019-620EA 187 USD
77019-620 77019-622 77019-624 77019-626
3-Bromodibenzothiophene 98%
3-Bromodibenzothiophene

Precaución: for Research Use Only

Formula: C₁₂H₇BrS
MDL Number: MFCD11052997
CAS Number: 97511-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR