(R)-tert-Butyl 2-amino-3-(3-bromophenyl)propanoate 98+%

Supplier: Ambeed
Warning

A839610-1G A839610-250MG
77019-616EA 237.65 USD
77019-616 77019-618
(R)-tert-Butyl 2-amino-3-(3-bromophenyl)propanoate 98+%
(R)-tert-Butyl 2-amino-3-(3-bromophenyl)propanoate

Precaución: for Research Use Only

Formula: C₁₃H₁₈BrNO₂
MW: 300.19 g/mol
MDL Number: MFCD07368810
CAS Number: 1241682-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR