1-p-Tolyl-1H-pyrazole-4-carbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 1-(p-Tolyl)-1H-pyrazole-4-carbaldehyde

A835763-1G A835763-250MG A835763-5G
77006-762EA 92 USD
77006-762 77006-764 77006-766
1-p-Tolyl-1H-pyrazole-4-carbaldehyde 98%
1-p-Tolyl-1H-pyrazole-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂O
MW: 186.22 g/mol
MDL Number: MFCD03426978
CAS Number: 337957-59-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR