4-Amino-2-methylbutan-2-ol 95%

Supplier: Ambeed
Danger

A834661-100MG A834661-1G A834661-250MG A834661-5G
76993-510EA 85.94 USD
76993-510 76993-512 76993-514 76993-516
4-Amino-2-methylbutan-2-ol 95%
4-Amino-2-methylbutan-2-ol

Precaución: for Research Use Only

Formula: C₅H₁₃NO
MW: 103.16 g/mol
MDL Number: MFCD00129174
CAS Number: 26734-08-7
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR