[4-Amino-2-(trifluoromethyl)phenyl](4-methyl-1-piperazinyl)methanone 97%

Supplier: Ambeed
Warning

A834574-1G A834574-250MG A834574-5G
77019-600EA 94 USD
77019-600 77019-602 77019-604
[4-Amino-2-(trifluoromethyl)phenyl](4-methyl-1-piperazinyl)methanone 97%
[4-Amino-2-(trifluoromethyl)phenyl](4-methyl-1-piperazinyl)methanone

Precaución: for Research Use Only

Formula: C₁₃H₁₆F₃N₃O
MDL Number: MFCD18911740
CAS Number: 853297-04-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR