6-Ethoxy-4-hydroxyquinoline 95%

Supplier: Ambeed
Danger

Synonyms: 6-Ethoxyquinolin-4-ol

A834531-100MG A834531-1G A834531-250MG A834531-5G
77036-592EA 7 USD
77036-592 77036-594 77036-596 77036-598
6-Ethoxy-4-hydroxyquinoline 95%
6-Ethoxy-4-hydroxyquinoline

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₂
MW: 189.211 g/mol
MDL Number: MFCD02332227
CAS Number: 303121-11-1
UN: 1759
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR