2,6-Dimethyl-4-fluorobenzonitrile 95%

Supplier: Ambeed
Danger

Synonyms: 4-Fluoro-2,6-dimethylbenzonitrile

A833849-100MG A833849-1G A833849-250MG A833849-5G
77019-592EA 15 USD
77019-592 77019-594 77019-596 77019-598
2,6-Dimethyl-4-fluorobenzonitrile 95%
2,6-Dimethyl-4-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₉H₈FN
MDL Number: MFCD04972839
CAS Number: 14659-61-1
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR