2-(3-Methoxyphenoxy)propionic acid 97%

Supplier: Ambeed
Warning

A833372-250MG A833372-1G A833372-5G
77006-760EA 10 USD
77006-760 77674-232 77674-234
2-(3-Methoxyphenoxy)propionic acid 97%
2-(3-Methoxyphenoxy)propionic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₄
MDL Number: MFCD01931912
CAS Number: 7309-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR