Ethyl 4′-hydroxy-4-biphenylcarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl-4'-hydroxy[1,1'-biphenyl]-4-carboxylate, Ethyl 4'-hydroxy-[1,1'-biphenyl]-4-carboxylate

A832139-1G A832139-250MG A832139-5G
77019-584EA 40.32 USD
77019-584 77019-586 77684-792
Ethyl 4′-hydroxy-4-biphenylcarboxylate 98%
Ethyl 4′-hydroxy-4-biphenylcarboxylate

Precaución: For research use only.

Formula: C₁₅H₁₄O₃
MW: 242.28 g/mol
MDL Number: MFCD07809563
CAS Number: 50670-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR