4-(2,5-Dimethyl-1H-pyrrol-1-yl)benzenecarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-(2,5-Dimethyl-1H-pyrrol-1-yl)benzoic acid

A829954-1G A829954-250MG A829954-5G
77036-578EA 67.9 USD
77036-578 77036-580 77036-582
4-(2,5-Dimethyl-1H-pyrrol-1-yl)benzenecarboxylic acid 98%
4-(2,5-Dimethyl-1H-pyrrol-1-yl)benzenecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO₂
MW: 215.25 g/mol
MDL Number: MFCD00453921
CAS Number: 15898-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR