Methyl 1-(4-fluorophenyl)cyclopropanecarboxylate 98%

Supplier: Ambeed
Warning

A829947-1G A829947-5G
77019-574EA 65.78 USD
77019-574 77684-790
Methyl 1-(4-fluorophenyl)cyclopropanecarboxylate 98%
Methyl 1-(4-fluorophenyl)cyclopropanecarboxylate

Precaución: For research use only.

Formula: C₁₁H₁₁FO₂
MW: 194.202 g/mol
MDL Number: MFCD18434442
CAS Number: 943111-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR