4-Hydroxy-6-methoxyquinoline-2-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A829903-250MG
A829903-100MG
A829903-1G
76993-496EA
36
USD
76993-496
77891-581
77891-582
4-Hydroxy-6-methoxyquinoline-2-carboxylic acid 95%
4-Hydroxy-6-methoxyquinoline-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₁H₉NO₄ |
MDL Number:
MFCD00047599 CAS Number: 91092-95-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...