Prazosin hydrochloride 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A829627-100G A829627-100MG A829627-25G A829627-5G A829627-1G A829627-250MG A829627-1MG
76993-488EA 241 USD
76993-488 76993-490 76993-492 76993-494 77614-482 77614-484 77684-786
Prazosin hydrochloride 95%, Ambeed.Inc
Prazosin hydrochloride

Precaución: For research use only.

Formula: C₁₉H₂₁N₅O₄
MDL Number: MFCD00058177
CAS Number: 19237-84-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR