Ethyl-3-oxo-3,4-dihydro-2-quinoxalinecarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-Oxo-3,4-dihydro-2-quinoxalinecarboxylate

A829598-1G A829598-250MG A829598-5G
77006-744EA 43.5 USD
77006-744 77006-746 77006-748
Ethyl-3-oxo-3,4-dihydro-2-quinoxalinecarboxylate 98%
Ethyl-3-oxo-3,4-dihydro-2-quinoxalinecarboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂O₃
MW: 218.21 g/mol
MDL Number: MFCD00459589
CAS Number: 36818-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR